missing translation for 'onlineSavingsMsg'
Learn More

Idoxuridine, TRC

Product Code. 30389874
Change view
Click to view available options
Quantity:
50 mg
2.5 g
5 g
Unit Size:
2.5g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389874 2.5 g 2.5g
30408737 50 mg 50mg
30406604 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389874 Supplier Toronto Research Chemicals Supplier No. TRCI2057502.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-42-2
Molecular Formula C9 H11 I N2 O5
Molecular Weight (g/mol) 354.1
Synonym Idoxuridine, Uridine, 2'-deoxy-5-iodo- (6CI, 7CI, 8CI, 9CI, ACI), 2'-Deoxy-5-iodouridine (ACI), (+)-5-Iodo-2'-deoxyuridine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-iodouracil, 5-Iodo-2'-deoxyuridine, 5-Iodo-2'-desoxyuridine, 5-Iododeoxyuridine, 5-Iodouracil deoxyriboside, 5IUDR, Allergan 211, Dendrid, Emanil, Herpe-Gel, Herpesil, Herpidu, Herplex, Idexur, Idoxene, Idoxuridin, IDU, Idu Oculos, Iducher, Idulea, IDUR, Iduridin, Iododeoxyuridine, Iododeozyuridine, IUDR, Joddeoxiuridin, Kerecid, MeSH ID: D007065, NSC 39661, Ophthalmadine, SKF 14287, Stoxil, Stoxip, Synmiol, Virudox
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(I)C(=O)NC2=O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C9H11IN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1
Formula Weight 353.9713
Chemical Name or Material Idoxuridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.