missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-45209
This item is not returnable.
View return policy
Description
IDO2 Polyclonal specifically detects IDO2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Peptide ELISA.
Specifications
| IDO2 | |
| Polyclonal | |
| Western Blot 1 - 3 ug/ml, Immunohistochemistry-Frozen 1:10 - 1:500, Peptide ELISA Detection limit 1:64000 | |
| NP_919270.2 | |
| IDO2 | |
| Peptide with sequence C-RDKTLESILHPR corresponding to C-Terminus according to NP_919270.2. | |
| 0.1 mg | |
| Primary | |
| This antibody is specific to the C-Terminus. Cross reacts with Human. Expected from sequence similarity: Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry (Frozen), ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| EC 1.13.11.-, IDO-2, INDOL1indoleamine-pyrrole 2,3 dioxygenase-like 1, indoleamine 2,3-dioxygenase 2, indoleamine 2,3-dioxygenase-like 1 protein, Indoleamine 2,3-dioxygenase-like protein 1, Indoleamine-pyrrole 2,3-dioxygenase-like protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 169355 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction