missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO2 Antibody (IDO2/2638), DyLight 550, Novus Biologicals™
Description
IDO2 Monoclonal antibody specifically detects IDO2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | IDO2 |
| Applications | ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | IDO2/2638 |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 1.13.11.-, IDO-2, INDOL1indoleamine-pyrrole 2,3 dioxygenase-like 1, indoleamine 2,3-dioxygenase 2, indoleamine 2,3-dioxygenase-like 1 protein, Indoleamine 2,3-dioxygenase-like protein 1, Indoleamine-pyrrole 2,3-dioxygenase-like protein 1 |
| Host Species | Mouse |
| Immunogen | Recombinant fragment (around aa 200-350) of human IDO2 protein (exact sequence is proprietary) (Uniprot: Q6ZQW0) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?