Learn More
Invitrogen™ IDO Monoclonal Antibody (eyedio), Alexa Fluor™ 700, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£260.00 - £552.00
Specifications
| Antigen | IDO |
|---|---|
| Clone | eyedio |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16676444
|
Invitrogen™
56-9477-41 |
25 Tests |
£260.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16686444
![]() |
Invitrogen™
56-9477-42 |
100 Tests |
£552.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This eyedio MAb reacts with human indoleamine-2, 3-dioxygenase (IDO, INDO, IDO1), an intracellular enzyme that catalyzes the degradation of tryptophan to kynurenines. IDO is expressed in a wide variety of tissues and cells, including macrophages, plasmacytoid dendritic cells, and several cell lines. IDO can be induced in many different cell types by IFN gamma or other inflammatory stimuli. Expression of IDO in antigen presenting cells and tumors is thought to mediate immune suppression through depletion of this essential amino acid and/or by the creation of tryptophan metabolites that cause apoptosis of T cells and induction of regulatory T cells.
IDO1 is an intracellular heme-containing enzyme that catalyzes the oxidative cleavage of the indole ring of several important regulatory molecules like tryptophan, serotonin, and melatonin. By doing this, IDO1 initiates the production of biologically active metabolites, commonly referred to as kynurenines. IDO1 is widely expressed in a variety of human tissues as well as in macrophages and dendritic cells (DCs). In inflammation, interferons (IFNs) act on specific receptors to trigger IDO1 induction. The production of IFN-gamma and induction of IDO1 represent important antimicrobial mechanisms. Degradation and depletion of tryptophan by IDO1 inhibits the growth of viruses, bacteria and parasites. Furthermore, IDO1 plays a complex and crucial role in immunoregulation during infection, pregnancy, autoimmunity, transplantation, and neoplasia.Specifications
| IDO | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| EC 1.13.11.52; IDO; IDO1; IDO-1; INDO; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; indoleamine-pyrrole 2,3-dioxygenase | |
| Ido1 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Ido1 |
| eyedio | |
| Flow Cytometry | |
| Alexa Fluor 700 | |
| Mouse | |
| Human | |
| P14902 | |
| 3620 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.