missing translation for 'onlineSavingsMsg'
Learn More

IDO-IN-7, MedChemExpress

Product Code. 16452097
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16452097

Brand: Medchem Express HY13983

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IDO-IN-7 (NLG-919 analogue) is a a potent IDO1 inhibitor with an IC50 of 38 nM.

IDO-IN-7 (NLG-919 analogue) is a a potent IDO1 inhibitor with an IC50 of 38 nM.

Specifications

Chemical Name or Material IDO-IN-7
CAS 1402836-58-1
Color White
Purity Grade Notes Research
Molecular Formula C18H22N2O
Quantity 50 mg
Synonym NLG-919 analogue GDC-0919 analogue
Solubility Information DMSO : 50 mg/mL (177.07 mM; Need ultrasonic) ∣Ethanol : 25 mg/mL (88.53 mM; Need ultrasonic)
SMILES OC(C1CCCCC1)CC(C2=C3C=CC=C2)N4C3=CN=C4
Molecular Weight (g/mol) 282.38
Formula Weight 282.38
Percent Purity 96.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.