missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IDI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100567
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total IDI1.
IDI1 encodes a peroxisomally-localized enzyme that catalyzes the interconversion of isopentenyl diphosphate (IPP) to its highly electrophilic isomer, dimethylallyl diphosphate (DMAPP), which are the substrates for the successive reaction that results in the synthesis of farnesyl diphosphate and, ultimately, cholesterol. It has been shown in peroxisomal deficiency diseases such as Zellweger syndrome and neonatal adrenoleukodystrophy that there is reduction in IPP isomerase activity.
Specifications
| IDI1 | |
| Polyclonal | |
| Unconjugated | |
| IDI1 | |
| 4832416K17Rik; IDI1; IPP isomerase 1; IPP1; IPPI1; isopentenyl diphosphate dimethylallyl diphosphate isomerase 1; isopentenyl pyrophosphate isomerase 1; isopentenyl-diphosphate delta isomerase; isopentenyl-diphosphate delta isomerase 1; isopentenyl-diphosphate Delta-isomerase 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 319554, 3422, 89784 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35760, P58044, Q13907 | |
| IDI1 | |
| A synthesized peptide derived from human IDI1(Accession Q13907), corresponding to amino acid residues L17-L67. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction