missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IDH3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575756
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the beta subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| IDH3B | |
| Polyclonal | |
| Unconjugated | |
| Idh3b | |
| C78231; DH3B; H-IDHB; H-IDHG; IDH3B; Isocitrate dehydrogenase [NAD] subunit beta, mitochondrial; isocitrate dehydrogenase 3 (NAD(+)) beta; isocitrate dehydrogenase 3 (NAD+) beta; isocitrate dehydrogenase 3 beta; isocitric dehydrogenase subunit beta; N14A tumor-related protein; NAD(+)-specific ICDH subunit beta; NAD+-specific isocitrate dehydrogenase b subunit; RP46 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 170718, 3420, 94173 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43837, Q68FX0 | |
| Idh3b | |
| Synthetic peptide corresponding to residues in Human IDH3B. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction