missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23254-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to IDH2.
Plays a role in intermediary metabolism and energy production. It may tightly associate or interact with the pyruvate dehydrogenase complex.Specifications
| IDH2 | |
| Polyclonal | |
| Unconjugated | |
| IDH2 | |
| ICD M, IDH, IDH2, IDHM, IDP, IDPM, mNADP IDH, NADP(+) specific ICDH, Oxalosuccinate decarboxylase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 269951, 3418, 361596 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P48735, P54071, P56574 | |
| Idh2 | |
| IDH2 Fusion Protein Ag19748 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction