missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IDH1 Polyclonal Antibody

Product Code. 12849122
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12849122 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12849122 Supplier Invitrogen™ Supplier No. PA520976

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is HepG2 cell lysate. PA5-20976 can be used with blocking peptide PEP-1090.

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP(+)-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP(+)-dependent isocitrate dehydrogenase found in the cytoplasm and peroxisomes. It contains the PTS-1 peroxisomal targeting signal sequence. The presence of this enzyme in peroxisomes suggests roles in the regeneration of NADPH for intraperoxisomal reductions, such as the conversion of 2, 4-dienoyl-CoAs to 3-enoyl-CoAs, as well as in peroxisomal reactions that consume 2-oxoglutarate, namely the alpha-hydroxylation of phytanic acid. The cytoplasmic enzyme serves a significant role in cytoplasmic NADPH production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDH1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene IDH1
Gene Accession No. O75874, O88844, P41562
Gene Alias AI314845; AI788952; cb876; corneal epithelial crystallin; cytosolic NADP+-dependent isocitrate dehydrogenase; cytosolic NADP-isocitrate dehydrogenase; cytosolic NADP-isocitrate dehydrongenase; E030024J03Rik; epididymis luminal protein 216; epididymis secretory protein Li 26; fm90e09; HEL-216; HEL-S-26; ICDH; Id-1; IDCD; IDH; Idh1; Idh-1; IDP; Idpc; im:7143416; isocitrate dehydrogenase (NAD(+)) IDH1; isocitrate dehydrogenase (NADP(+)) 1; isocitrate dehydrogenase (NADP(+)) 1, cytosolic; Isocitrate dehydrogenase [NAD] subunit 1, mitochondrial; Isocitrate dehydrogenase [NADP] cytoplasmic; isocitrate dehydrogenase 1 (NADP+); isocitrate dehydrogenase 1 (NADP+), soluble; Isocitrate dehydrogenase 1, soluble; Isocitric dehydrogenase; N2690; NAD(+)-specific ICDH; NADP(+)-specific ICDH; NADP+-specific ICDH; NADP-cICDH; NADP-dependent isocitrate dehydrogenase, cytosolic; NADP-dependent isocitrate dehydrogenase, peroxisomal; OTTHUMP00000206464; OTTHUMP00000206465; OTTHUMP00000206466; oxalosuccinate decarboxylase; PICD; wu:fm90e09; YNL037C
Gene Symbols IDH1
Host Species Rabbit
Immunogen A 13 amino acid peptide near the carboxy terminus of human IDH1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15926, 24479, 3417
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.