missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH1 Mouse anti-Human, Monkey, Mouse, Clone:7G8A1, Abnova™
Mouse monoclonal antibody raised against recombinant human IDH1.
Brand: Abnova MAB17855.100ug
This item is not returnable.
View return policy
Description
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP(+)-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP(+)-dependent isocitrate dehydrogenase found in the cytoplasm and peroxisomes. It contains the PTS-1 peroxisomal targeting signal sequence. The presence of this enzyme in peroxisomes suggests roles in the regeneration of NADPH for intraperoxisomal reductions, such as the conversion of 2, 4-dienoyl-CoAs to 3-enoyl-CoAs, as well as in peroxisomal reactions that consume 2-oxoglutarate, namely the alpha-hydroxylation of phytanic acid. The cytoplasmic enzyme serves a significant role in cytoplasmic NADPH production. [provided by RefSeq]Specifications
| IDH1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-400) Immunofluorescence (1:50-250) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| IDH1 | |
| Mouse | |
| 100 μg | |
| Human, Monkey, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Western Blot | |
| 7G8A1 | |
| isocitrate dehydrogenase 1 (NADP+), soluble | |
| In PBS (0.05% sodium azide) | |
| IDCD, IDH, IDP, IDPC, PICD | |
| Recombinant protein corresponding to amino acids 156-298 of human IDH1 from E. coli. | |
| 3417 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction