missing translation for 'onlineSavingsMsg'
Learn More

Idebenone, TRC

Product Code. 30133161
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133161 100 mg 100mg
30004572 10 mg 10mg
30007084 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133161 Supplier Toronto Research Chemicals Supplier No. TRCI170050100MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58186-27-9
Molecular Formula C19 H30 O5
Molecular Weight (g/mol) 338.44
Synonym 2,5-Cyclohexadiene-1,4-dione, 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methyl-, 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione, 2,3-Dimethoxy-5-methyl-6-(10-hydroxydecyl)-1,4-benzoquinone, 2,3-Dimethoxy-6-(10-hydroxydecyl)-5-methyl-1,4-benzoquinone, 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione, Avan, CV 2619, Daruma, Idebenone, Mnesis, Raxone
IUPAC Name 2-(10-hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
SMILES COC1=C(OC)C(=O)C(=C(C)C1=O)CCCCCCCCCCO

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H30O5/c1-14-15(12-10-8-6-4-5-7-9-11-13-20)17(22)19(24-3)18(23-2)16(14)21/h20H,4-13H2,1-3H3
Formula Weight 338.2093
Chemical Name or Material Idebenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.