missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies IDE2
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- Inducer of definitive endoderm 2 (IDE2) induces differentiation of mouse or human pluripotent stem cells (PSCs) by activating SMAD2 phosphorylation and NODAL expression (Borowiak et al.) At EC50 = 223nM, SOX17 expression was induced in mouse ES cells
- Induces definitive endoderm from mouse or human ES cells in the absence of Activin A, NODAL, or feeder cells (Borowiak et al.)
Specifications
Specifications
| CAS | 1136466-93-7 |
| Quantity | 1 mg |
| Molecular Formula | C12H20N2O3 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction