missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies IDE2
Description
- Inducer of definitive endoderm 2 (IDE2) induces differentiation of mouse or human pluripotent stem cells (PSCs) by activating SMAD2 phosphorylation and NODAL expression (Borowiak et al.) At EC50 = 223nM, SOX17 expression was induced in mouse ES cells
- Induces definitive endoderm from mouse or human ES cells in the absence of Activin A, NODAL, or feeder cells (Borowiak et al.)
Specifications
Specifications
| CAS | 1136466-93-7 |
| Quantity | 5 mg |
| Molecular Formula | C12H20N2O3 |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?