missing translation for 'onlineSavingsMsg'
Learn More

Idasanutlin, MedChemExpress

Product Code. 16439287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439287 5 mg 5mg
16442817 50 mg 50mg
16436738 10 mg 10mg
16406876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16439287 Supplier Medchem Express Supplier No. HY15676
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Idasanutlin (RG7388) is a potent and selective MDM2 antagonist, inhibiting p53-MDM2 binding, with an IC50 of 6 nM.

Idasanutlin (RG7388) is a potent and selective MDM2 antagonist, inhibiting p53-MDM2 binding, with an IC50 of 6 nM.

Specifications

Chemical Name or Material Idasanutlin
CAS 1229705-06-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H29Cl2F2N3O4
Quantity 5 mg
Synonym RG7388
Solubility Information DMSO : ≥ 45 mg/mL (73.00 mM)
SMILES O=C(O)C1=CC(OC)=C(NC([C@H]2[C@H](C3=C(F)C(Cl)=CC=C3)[C@](C4=CC=C(Cl)C=C4F)(C#N)[C@H](CC(C)(C)C)N2)=O)C=C1
Molecular Weight (g/mol) 616.48
Formula Weight 616.48
Percent Purity 97.15%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.