missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ID4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101114
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ID4.
ID4 is transcription factors containing a basic helix-loop-helix (bHLH) motif regulate expression of tissue-specific genes in a number of mammalian and insect systems. DNA-binding activity of the bHLH proteins is dependent on formation of homo- and/or heterodimers. Dominant-negative HLH proteins encoded by Id-related genes, such as ID4, also contain the HLH-dimerization domain but lack the DNA-binding basic domain. Consequently, Id proteins inhibit binding to DNA and transcriptional transactivation by heterodimerization with bHLH proteins.
Specifications
| ID4 | |
| Polyclonal | |
| Unconjugated | |
| Id4 | |
| bHLHb27; cb547; Class B basic helix-loop-helix protein 27; DNA-binding protein inhibitor ID-4; fb77d04; hypothetical protein LOC664761; ID4; Id-4; Idb4; Inhibitor of differentiation 4; inhibitor of DNA binding 4; inhibitor of DNA binding 4, dominant negative helix-loop-helix protein; inhibitor of DNA binding 4, HLH protein; sb:cb547; wu:fb77d04; zgc:123214 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15904, 291023, 3400 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P41139, P47928 | |
| Id4 | |
| A synthesized peptide derived from human ID4(Accession P47928), corresponding to amino acid residues M1-G36. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction