missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ID4 Antibody (3F11), Novus Biologicals™

Product Code. 18346608 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346608 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346608 Supplier Novus Biologicals™ Supplier No. H00003400M13

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ID4 Monoclonal antibody specifically detects ID4 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ID4
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 3F11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001537.1
Gene Alias bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein
Host Species Mouse
Immunogen ID4 (NP_001537.1, 1 a.a. ∽ 161 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKAVSPVRPSGRKAPSGCGGGELALRCLAEHGHSLGGSAAAAAAAAAARCKAAEAAADEPALCLQCDMNDCYSRLRRLVPTIPPNKKVSKVEILQHVIDYILDLQLALETHPALLRQPPPPAPPHHPAGTCPAAPPRTPLTALNTDPAGAVNKQGDSILCR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3400
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.