missing translation for 'onlineSavingsMsg'
Learn More

ID3 Antibody, Novus Biologicals™

Product Code. 18109288 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18109288 0.1 mL 0.1mL
18686655 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18109288 Supplier Novus Biologicals Supplier No. NBP247423

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ID3 Polyclonal specifically detects ID3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ID3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias BHLHB25, bHLHb251R21, Class B basic helix-loop-helix protein 25, DNA-binding protein inhibitor ID-3, HEIR1, HEIR-1, Helix-loop-helix protein HEIR-1, ID-like protein inhibitor HLH 1R21, Inhibitor of DNA binding 3, inhibitor of DNA binding 3, dominant negative helix-loop-helix protein
Gene Symbols ID3
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: VPRGTQLSQVEILQRVIDYILDLQVVLAEPAPGPPDGPHLPIQTAELAPELVISND
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3399
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.