missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID2 Mouse anti-Human, Clone:4E12G5, Abnova™
Mouse monoclonal antibody raised against recombinant human ID2.
Brand: Abnova MAB17798.100ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the inhibitor of DNA binding (ID) family, members of which are transcriptional regulators that contain a helix-loop-helix (HLH) domain but not a basic domain. Members of the ID family inhibit the functions of basic helix-loop-helix transcription factors in a dominant-negative manner by suppressing their heterodimerization partners through the HLH domains. This protein may play a role in negatively regulating cell differentiation. A pseudogene has been identified for this gene. [provided by RefSeq]Specifications
| ID2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunocytochemistry Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
| ID2 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 4E12G5 | |
| inhibitor of DNA binding 2, dominant negative helix-loop-helix protein | |
| In PBS (0.05% sodium azide) | |
| GIG8, ID2A, ID2H, MGC26389, bHLHb26 | |
| Recombinant protein corresponding to amino acids 1-134 of human ID2 from E. coli. | |
| 3398 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction