Learn More
Invitrogen™ ID2 Monoclonal Antibody (ILCID2), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 48947582
Description
Description: This ILCID2 monoclonal antibody recognizes mouse and human ID2. This ILCID2 antibody requires Foxp3/Transcription Factor Staining Buffer set and will perform poorly when used with standard IC Fixation & Permeabilization Buffer Set and protocol. Applications Reported: This ILCID2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ILCID2 antibody has been tested by flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One step protocol for (nuclear) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative for Pacific Blue™. eFluor 450 emits at 446 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser
The inhibitor of DNA binding 2 (ID2) protein is essential for promoting the development of a number of cell types including natural killer cell, erythroid precursors, as well as, CD8+ and CD4+ T-cell. A lack of ID2 promotes B-cell development. The chief role of this protein is to associate with ubiquitously-expressed E proteins, and other proteins of the basic helix-loop-helix (HLH) family, preventing them from binding to DNA and activating transcription. Activation of ID2 is critical for the development of the natural killer cell lineage.
Specifications
| ID2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P41136, Q02363 | |
| ID2 | |
| Purified recombinant fragment of human ID2, corresponding to amino acids 1-134 expressed in E. coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| ILCID2 | |
| eFluor 450 | |
| ID2 | |
| Ac2-300; AI255428; bHLHb26; C78922; cell growth-inhibiting gene 8; class B basic helix-loop-helix protein 26; DNA-binding protein inhibitor ID2; DNA-binding protein inhibitor ID-2; GIG8; helix-loop-helix protein ID2; Id2; Id-2; ID2A; ID2H; Idb2; Inhibitor of differentiation 2; inhibitor of DNA binding 2; inhibitor of DNA binding 2, dominant negative helix-loop-helix protein; inhibitor of DNA binding 2, HLH protein; MGC26389; OTTHUMP00000140258; OTTHUMP00000200297 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 15902, 3398 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.