missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99044AF700
This item is not returnable.
View return policy
Description
ID2 Polyclonal specifically detects ID2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ID2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 700 | |
| BHLHB26, bHLHb26cell growth-inhibiting gene 8, Class B basic helix-loop-helix protein 26, DNA-binding protein inhibitor ID2, DNA-binding protein inhibitor ID-2, GIG8, helix-loop-helix protein ID2, ID2A, ID2H, inhibitor of differentiation 2, Inhibitor of DNA binding 2, inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, MGC26389 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ID2. | |
| 0.1 mL | |
| Cancer, Innate Immunity | |
| 3398 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction