missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ID2 Antibody (A4-D4), Novus Biologicals™
Mouse Monoclonal Antibody
£167.00 - £524.00
Specifications
| Antigen | ID2 |
|---|---|
| Clone | A4-D4 |
| Concentration | 2 mg/mL |
| Dilution | Western Blot 1:1000-1:10000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056884
|
Novus Biologicals™
NBP3-30992 |
100 μL |
£524.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30057160
|
Novus Biologicals™
NBP3-30992-25ul |
25 μL |
£167.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ID2 Monoclonal antibody specifically detects ID2 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ ImmunofluorescenceSpecifications
| ID2 | |
| 2 mg/mL | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Innate Immunity | |
| PBS, pH 7.4, 40% Glycerol, 0.2% BSA | |
| 3398 | |
| IgG2b | |
| Protein A purified |
| A4-D4 | |
| Western Blot 1:1000-1:10000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| BHLHB26, bHLHb26cell growth-inhibiting gene 8, Class B basic helix-loop-helix protein 26, DNA-binding protein inhibitor ID2, DNA-binding protein inhibitor ID-2, GIG8, helix-loop-helix protein ID2, ID2A, ID2H, inhibitor of differentiation 2, Inhibitor of DNA binding 2, inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, MGC26389 | |
| Recombinant protein with Human ID2 aa 1- 80 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto