missing translation for 'onlineSavingsMsg'
Learn More

ID1 Antibody (PCRP-ID1-2F11), DyLight 755, Novus Biologicals™

Product Code. 18391976 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391976 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391976 Supplier Novus Biologicals Supplier No. NBP314075IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ID1 Monoclonal antibody specifically detects ID1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ID1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-ID1-2F11
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias BHLHB24, bHLHb24DNA-binding protein inhibitor ID-1, Class B basic helix-loop-helix protein 24, dJ857M17.1.2, dJ857M17.1.2 (inhibitor of DNA binding 1, dominant negative helix-loop-helixprotein), ID, inhibitor of differentiation 1, Inhibitor of DNA binding 1, inhibitor of DNA binding 1, dominant negative helix-loop-helix protein
Host Species Mouse
Immunogen Recombinant fragment (aa 5-150) of human ID1 (Uniprot: P41134)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3397
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.