missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145150
This item is not returnable.
View return policy
Description
Immunogen sequence: ITEASQTPKE PTKEDVKLHR IRIENMNRER LRQKRIHSAV KTSRRVDMD Antigen sequence identity: Mouse 73%; Rat 71%.
Essential peptidyl-tRNA hydrolase component of the mitochondrial large ribosomal subunit. Acts as a codon-independent translation release factor that has lost all stop codon specificity and directs the termination of translation in mitochondrion, possibly in case of abortive elongation. May be involved in the hydrolysis of peptidyl-tRNAs that have been prematurely terminated and thus in the recycling of stalled mitochondrial ribosomes.
Specifications
| ICT1 | |
| Polyclonal | |
| Unconjugated | |
| MRPL58 | |
| 1110001A02Rik; 1110002E03Rik; 39S ribosomal protein L58, mitochondrial; Digestion substraction 1; DS1; DS-1; Ict1; immature colon carcinoma transcript 1; immature colon carcinoma transcript 1 protein; Immature colon carcinoma transcript 1 protein homolog; Mitochondrial large ribosomal subunit protein mL62; mitochondrial ribosomal protein L58; MRPL58; MRP-L58; Peptidyl-tRNA hydrolase ICT1, mitochondrial | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3396 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14197 | |
| MRPL58 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 158-206. Recombinant protein control fragment (Product #RP-110113). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction