missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICT Antibody (1E9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42633B
This item is not returnable.
View return policy
Description
ICT Monoclonal specifically detects ICT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| ICT | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| MRPL58 | |
| Recombinant human ICT1 (30-206aa) purified from E. coli | |
| 100 μL | |
| Cancer | |
| 3396 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA | |
| 1000000000 | |
| Western Blot, Flow Cytometry, ELISA | |
| Digestion substraction 1, DS-1DS1, EC 3.1.1.29, immature colon carcinoma transcript 1, Immature colon carcinoma transcript 1 protein, peptidyl-tRNA hydrolase ICT1, mitochondrial | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction