missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ICSP Polyclonal Antibody
GREENER_CHOICE

Product Code. 17960473
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17960473 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17960473 Supplier Invitrogen™ Supplier No. PA5144411

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ICSP Polyclonal Antibody for Western Blot, ELISA

Protease responsible for the cleavage of IcsA between 'Arg-758' and 'Arg-759', removing the entire alpha domain from IscA localized on the bacterial surface. This proteolytic activity contributes to the maintenance of a tight polar cap of IcsA, which is important to Shigella actin-based motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICSP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene icsP
Gene Accession No. O33641
Gene Alias sopA
Gene Symbols icsP
Host Species Rabbit
Immunogen Recombinant Shigella flexneri ICSP protein (21-315aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1238162
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.