missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICS Interference A: Fe @ 2000; Al, Ca, Mg @ 5000 μg/mL in 20% HCl, VHG™
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Description
ICS Interference A: Fe @ 2000; Al, Ca, Mg @ 5000 μg/mL in 20% HCl
Specifications
Specifications
| Chemical Name or Material | Aluminium/Calcium/Iron/Magnesium |
| CAS | 7429-90-5, 7440-70-2, 7439-89-6, 7439-95-4 |
| Methods | ICP |
| Analyte or Component Names | Aluminium/Calcium/Iron/Magnesium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HCl |
| Quantity | 250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction