missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICS Analytes B: Pb, Se @ 5; As, Tl @ 10; Ag @ 20; Ba, Be, Co, Cr, Cu, Mn, V @ 50; Sb @ 60; Cd, Ni, Zn @ 100 μg/mL in 5% HNO3, tr. Tartaric Acid, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
ICS Analytes B: Pb, Se @ 5; As, Tl @ 10; Ag @ 20; Ba, Be, Co, Cr, Cu, Mn, V @ 50; Sb @ 60; Cd, Ni, Zn @ 100 μg/mL in 5% HNO3, tr. Tartaric Acid
Specifications
Specifications
| Chemical Name or Material | Lead/Selenium/Arsenic/Thallium/Silver/Barium/Beryllium/Chromium/Cobalt/Copper/Manganese/Vanadium/Antimony/Cadmium/Nickel/Zinc |
| CAS | 7439-92-1, 7782-49-2, 7440-38-2, 7440-28-0, 7440-22-4, 7440-39-3 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Lead/Selenium/Arsenic/Thallium/Silver/Barium/Beryllium/Chromium/Cobalt/Copper/Manganese/Vanadium/Antimony/Cadmium/Nickel/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Lead 5 μg/mL/Selenium 5 μg/mL/Arsenic 10 μg/mL/Thallium 10 μg/mL/Silver 20 μg/mL/Barium 50 μg/mL/Beryllium 50 μg/mL/Chromium 50 μg/mL/Cobalt 50 μg/mL/Copper 50 μg/mL/Manganese 50 μg/mL/Vanadium 50 μg/mL/Antimony 60 μg/mL/Cadmium 100 μg/mL/Nickel 100 μg/mL/Zinc 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction