Learn More
Icrucumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA541931
Additional Details : Weight : 0.01000kg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Icrucumab is under investigation in clinical trial NCT01111604 (A Study of IMC-1121B or IMC-18F1 in Colorectal Cancer).Specifications
Icrucumab | |
Recombinant Monoclonal | |
Unconjugated | |
18F1; IMC-18F1; LY3012212 | |
Human FLT1/ VEGFR-1 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 κ |