missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iCRT-14, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | iCRT-14 |
| CAS | 677331-12-3 |
| Molecular Formula | C21H17N3O2S |
| Quantity | 10 mg |
| Synonym | 5-[[2,5-Dimethyl-1-(3-pyridinyl)-1H-pyrrol-3-yl]methylene]-3-phenyl-2,4-thiazolidinedione, iCRT 14, iCRT14 |
| InChI Formula | InChI=1S/C21H17N3O2S/c1-14-11-16(15(2)23(14)18-9-6-10-22-13-18)12-19-20(25)24(21(26)27-19)17-7-4-3-5-8-17/h3-13H,1-2H3/b19-12- |
| SMILES | Cc1cc(\C=C\2/SC(=O)N(C2=O)c3ccccc3)c(C)n1c4cccnc4 |
| Molecular Weight (g/mol) | 375.44 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction