missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ iCRT 14
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent inhibitor of β-catenin-responsive transcription (CRT) (IC50 = 40.3 nM in assays of Wnt pathway activity). Thought to directly influence the interaction between β-catenin and TCF4. Induces marked G0/G1 cell cycle arrest in HCT-116 and HT29 cell lines.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 375.44 |
| Purity | >99% |
| Molecular Weight (g/mol) | 375.446 |
| Chemical Name or Material | iCRT 14 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction