missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICP-OES VIS Wave Cal Solution: K @ 50 μg/mL; La, Li, Mn, Na, Sr @ 10 μg/mL; Ba, Ca @ 1 μg/mL in 2% HNO3, VHG™
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Description
ICP-OES VIS Wave Cal Solution: K @ 50 μg/mL; La, Li, Mn, Na, Sr @ 10 μg/mL; Ba, Ca @ 1 μg/mL in 2% HNO3
Specifications
Specifications
| Chemical Name or Material | Barium/Calcium/Lanthanum/Lithium/Manganese/Sodium/Strontium/Potassium |
| CAS | 7440-39-3, 7440-70-2, 7439-91-0, 7439-93-2, 7439-96-5, 7440-23-5 |
| Analyte or Component Names | Barium/Calcium/Lanthanum/Lithium/Manganese/Sodium/Strontium/Potassium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 250 mL |
| Concentration or Composition (by Analyte or Components) | Barium 1 μg/mL/Calcium 1 μg/mL/Lanthanum 10 μg/mL/Lithium 10 μg/mL/Manganese 10 μg/mL/Sodium 10 μg/mL/Strontium 10 μg/mL/Potassium 50 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction