missing translation for 'onlineSavingsMsg'
Learn More

ICP-OES UV Wave Cal Solution: K, P, S @ 100 μg/mL; As, La, Li, Mn, Mo, Na, Ni, Sc @ 20 μg/mL; Ca @ 1 μg/mL in 5% HCl, VHG™

Product Code. 30003084
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003084 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003084 Supplier VHG Labs Supplier No. VHGISUPEUVW500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-OES UV Wave Cal Solution: K, P, S @ 100 μg/mL; As, La, Li, Mn, Mo, Na, Ni, Sc @ 20 μg/mL; Ca @ 1 μg/mL in 5% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium/Arsenic/Lanthanum/Lithium/Manganese/Molybdenum/Nickel/Scandium/Sodium/Phosphorus/Potassium
CAS 7440-70-2, 7440-38-2, 7439-91-0, 7439-93-2, 7439-96-5, 7439-98-7
Analyte or Component Names Calcium/Arsenic/Lanthanum/Lithium/Manganese/Molybdenum/Nickel/Scandium/Sodium/Phosphorus/Potassium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HCl
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Calcium 1 μg/mL/Arsenic 20 μg/mL/Lanthanum 20 μg/mL/Lithium 20 μg/mL/Manganese 20 μg/mL/Molybdenum 20 μg/mL/Nickel 20 μg/mL/Scandium 20 μg/mL/Sodium 20 μg/mL/Phosphorus 100 μg/mL/Potassium 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.