missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICP-MS Tuning Solution: 7Li, Be, Mg, Co, In, Ba, Ce, Pb, Bi, U @10 ug/L in 2% HNO3, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
ICP-MS Tuning Solution: 7Li, Be, Mg, Co, In, Ba, Ce, Pb, Bi, U @10 ug/L in 2% HNO3
Specifications
Specifications
| Chemical Name or Material | Beryllium/Cobalt/Indium/Lithium/Barium/Bismuth/Cerium/Uranium/Lead/Magnesium |
| CAS | 7440-41-7, 7440-48-4, 7440-74-6, 7439-93-2, 7440-39-3, 7440-69-9 |
| Methods | ICP-MS |
| Analyte or Component Names | Beryllium/Cobalt/Indium/Lithium/Barium/Bismuth/Cerium/Uranium/Lead/Magnesium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Beryllium 9.99 μg/L/Cobalt 10 μg/L/Indium 10.01 μg/L/Lithium 10.02 μg/L/Barium 10.03 μg/L/Bismuth 10.03 μg/L/Cerium 10.03 μg/L/Uranium 10.03 μg/L/Lead 10.06 μg/L/Magnesium 10.06 μg/L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction