missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICP-MS Tuning/Mass Calibration Multi-Element Mix 3 (concentrate): 7Li, Be, Mg, Co, Y, In, Ba, Ce, Tb, Pb, U @ 10 ug/mL in 5% HNO3, VHG™
Description
ICP-MS Tuning/Mass Calibration Multi-Element Mix 3 (concentrate): 7Li, Be, Mg, Co, Y, In, Ba, Ce, Tb, Pb, U @ 10 ug/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Cobalt/Indium/Lead/Lithium/Uranium/Beryllium/Cerium/Barium/Magnesium/Yttrium/Terbium |
| CAS | 7440-48-4, 7440-74-6, 7439-92-1, 7439-93-2, 7440-61-1, 7440-41-7 |
| Methods | ICP-MS |
| Analyte or Component Names | Cobalt/Indium/Lead/Lithium/Uranium/Beryllium/Cerium/Barium/Magnesium/Yttrium/Terbium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Cobalt 10 μg/mL/Indium 10 μg/mL/Lead 10 μg/mL/Lithium 10 μg/mL/Uranium 10 μg/mL/Beryllium 10.01 μg/mL/Cerium 10.01 μg/mL/Barium 10.02 μg/mL/Magnesium 10.02 μg/mL/Yttrium 10.02 μg/mL/Terbium 10.07 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?