missing translation for 'onlineSavingsMsg'
Learn More

ICP-MS Tuning/Mass Calibration Multi-Element Mix 2 (concentrate): Be, Mg, Co, In, Ce, Pb @ 10 ug/mL in 1% HNO3, VHG™

Product Code. 17978939
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 17978939

Brand: VHG Labs VHGLMSTNG2Z100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-MS Tuning/Mass Calibration Multi-Element Mix 2 (concentrate): Be, Mg, Co, In, Ce, Pb @ 10 ug/mL in 1% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beryllium/Cerium/Cobalt/Indium/Lead/Magnesium
CAS 7440-41-7, 7440-45-1, 7440-48-4, 7440-74-6, 7439-92-1, 7439-95-4
Methods ICP-MS
Analyte or Component Names Beryllium/Cerium/Cobalt/Indium/Lead/Magnesium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Beryllium 10 μg/mL/Cerium 10 μg/mL/Cobalt 10 μg/mL/Indium 10 μg/mL/Lead 10 μg/mL/Magnesium 10 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.