missing translation for 'onlineSavingsMsg'
Learn More

ICP-MS Tuning/Mass Calibration Multi-Element Mix 1A (Concentrate): 7Li, Co, Y, Ce, Tl @ 10 ug/mL in 1% HNO3, 0.5% HCl, VHG™

Product Code. 30003094
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003094 500 mL 500mL
30003105 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003094 Supplier VHG Labs Supplier No. VHGLMSTNG5CONC500
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-MS Tuning/Mass Calibration Multi-Element Mix 1A (Concentrate): 7Li, Co, Y, Ce, Tl @ 10 ug/mL in 1% HNO3, 0.5% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Yttrium/Cerium/Cobalt/Lithium/Thallium
CAS 7440-65-5, 7440-45-1, 7440-48-4, 7439-93-2, 7440-28-0
Methods ICP-MS
Analyte or Component Names Yttrium/Cerium/Cobalt/Lithium/Thallium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Yttrium 9.99 μg/mL/Cerium 10.01 μg/mL/Cobalt 10.02 μg/mL/Lithium 10.02 μg/mL/Thallium 10.04 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.