missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICP-MS P/A Tuning Solution 1: As, Be, Cd, Zn @ 20μg/mL; Mg, Ni, Pb @ 10 μg/mL; Al, Ba, Bi, Co, Cr, Cu, In, 6Li, Lu, Mn, Na, Sc, Sr, Th, Tl, U, V @ 5μg/mL; Y, Yb @ 2.5 μg/mL in 2% HNO3, VHG™
Description
ICP-MS P/A Tuning Solution 1: As, Be, Cd, Zn @ 20μg/mL; Mg, Ni, Pb @ 10 μg/mL; Al, Ba, Bi, Co, Cr, Cu, In, 6Li, Lu, Mn, Na, Sc, Sr, Th, Tl, U, V @ 5μg/mL; Y, Yb @ 2.5 μg/mL in 2% HNO3
Specifications
Specifications
| Chemical Name or Material | Ytterbium/Yttrium/Aluminium/Barium/Bismuth/Chromium/Cobalt/Copper/Indium/Lithium 6/Lutetium/Manganese/Scandium/Sodium/Strontium/Thallium/Thorium/Uranium/Vanadium/Lead/Magnesium/Nickel/Arsenic/Beryllium/Cadmium/Zinc |
| CAS | 7440-64-4, 7440-65-5, 7429-90-5, 7440-39-3, 7440-69-9, 7440-47-3 |
| Analyte or Component Names | Ytterbium/Yttrium/Aluminium/Barium/Bismuth/Chromium/Cobalt/Copper/Indium/Lithium 6/Lutetium/Manganese/Scandium/Sodium/Strontium/Thallium/Thorium/Uranium/Vanadium/Lead/Magnesium/Nickel/Arsenic/Beryllium/Cadmium/Zinc |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Ytterbium 2.5 μg/mL/Yttrium 2.5 μg/mL/Aluminium 5 μg/mL/Barium 5 μg/mL/Bismuth 5 μg/mL/Chromium 5 μg/mL/Cobalt 5 μg/mL/Copper 5 μg/mL/Indium 5 μg/mL/Lithium 6 5 μg/mL/Lutetium 5 μg/mL/Manganese 5 μg/mL/Scandium 5 μg/mL/Sodium 5 μg/mL/Strontium 5 μg/mL/Thallium 5 μg/mL/Thorium 5 μg/mL/Uranium 5 μg/mL/Vanadium 5 μg/mL/Lead 10 μg/mL/Magnesium 10 μg/mL/Nickel 10 μg/mL/Arsenic 20 μg/mL/Beryllium 20 μg/mL/Cadmium 20 μg/mL/Zinc 20 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?