missing translation for 'onlineSavingsMsg'
Learn More

ICP-MS P/A Tuning Mix 1: Y, Tb @ 2.5ug/mL; 6Li, Na, Al, Sc, Ti, V, Cr, Mn, Co, Cu, Sr, In, Ba, Lu, Ir, Bi, Tl, Th, U @ 5ug/mL; Mg, Ni, Ge, Mo, Ru, Pd, Sn, Sb, Pb @ 10ug/mL; Be, Zn, As, Cd @ 20 ug/mL in 20% HCl, tr. HF, VHG™

Product Code. 30003012
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003012 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30003012 Supplier VHG Labs Supplier No. VHGLDPA1100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-MS P/A Tuning Mix 1: Y, Tb @ 2.5ug/mL; 6Li, Na, Al, Sc, Ti, V, Cr, Mn, Co, Cu, Sr, In, Ba, Lu, Ir, Bi, Tl, Th, U @ 5ug/mL; Mg, Ni, Ge, Mo, Ru, Pd, Sn, Sb, Pb @ 10ug/mL; Be, Zn, As, Cd @ 20 ug/mL in 20% HCl, tr. HF
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Terbium/Yttrium/Aluminium/Barium/Bismuth/Chromium/Cobalt/Copper/Indium/Iridium/Lithium 6/Lutetium/Manganese/Scandium/Sodium/Strontium/Thallium/Thorium/Titanium/Uranium/Vanadium/Antimony/Germanium/Lead/Magnesium/Molybdenum/Nickel/Palladium/Tin/Arsenic/Beryllium/Cadmium/Zinc
CAS 7440-27-9, 7440-65-5, 7429-90-5, 7440-39-3, 7440-69-9, 7440-47-3
Methods ICP-MS
Analyte or Component Names Terbium/Yttrium/Aluminium/Barium/Bismuth/Chromium/Cobalt/Copper/Indium/Iridium/Lithium 6/Lutetium/Manganese/Scandium/Sodium/Strontium/Thallium/Thorium/Titanium/Uranium/Vanadium/Antimony/Germanium/Lead/Magnesium/Molybdenum/Nickel/Palladium/Tin/Arsenic/Beryllium/Cadmium/Zinc
Shipping Condition Room Temperature
Solvent or Matrix Dilute HCl
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Terbium 2.5 μg/mL/Yttrium 2.5 μg/mL/Aluminium 5 μg/mL/Barium 5 μg/mL/Bismuth 5 μg/mL/Chromium 5 μg/mL/Cobalt 5 μg/mL/Copper 5 μg/mL/Indium 5 μg/mL/Iridium 5 μg/mL/Lithium 6 5 μg/mL/Lutetium 5 μg/mL/Manganese 5 μg/mL/Scandium 5 μg/mL/Sodium 5 μg/mL/Strontium 5 μg/mL/Thallium 5 μg/mL/Thorium 5 μg/mL/Titanium 5 μg/mL/Uranium 5 μg/mL/Vanadium 5 μg/mL/Antimony 10 μg/mL/Germanium 10 μg/mL/Lead 10 μg/mL/Magnesium 10 μg/mL/Molybdenum 10 μg/mL/Nickel 10 μg/mL/Palladium 10 μg/mL/Tin 10 μg/mL/Arsenic 20 μg/mL/Beryllium 20 μg/mL/Cadmium 20 μg/mL/Zinc 20 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.