missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICP-MS Internal Standard Solution: Bi, Ge, In, Li-6, Rh, Sc, Tb, Y @ 100ug/mL in 5% HNO3, tr. F-, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
ICP-MS Internal Standard Solution: Bi, Ge, In, Li-6, Rh, Sc, Tb, Y @ 100ug/mL in 5% HNO3, tr. F-
Specifications
Specifications
| Chemical Name or Material | Bismuth/Germanium/Indium/Lithium 6/Rhodium/Scandium/Terbium/Yttrium |
| CAS | 7440-69-9, 7440-56-4, 7440-74-6, 14258-72-1, 7440-16-6, 7440-20-2 |
| Analyte or Component Names | Bismuth/Germanium/Indium/Lithium 6/Rhodium/Scandium/Terbium/Yttrium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Bismuth 100 μg/mL/Germanium 100 μg/mL/Indium 100 μg/mL/Lithium 6 100 μg/mL/Rhodium 100 μg/mL/Scandium 100 μg/mL/Terbium 100 μg/mL/Yttrium 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction