missing translation for 'onlineSavingsMsg'
Learn More

Icotinib, MedChemExpress

Product Code. 16496338
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496338

Brand: Medchem Express HY15164A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Icotinib (BPI-2009) is a potent and specific EGFR inhibitor with an IC50 of 5 nM; also inhibits mutant EGFRL858R, EGFRL858R/T790M, EGFRT790M and EGFRL861Q.

Icotinib (BPI-2009) is a potent and specific EGFR inhibitor with an IC50 of 5 nM

Specifications

Chemical Name or Material Icotinib
CAS 610798-31-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H21N3O4
Quantity 10 mg
Synonym BPI-2009
Solubility Information DMSO : ≥ 155 mg/mL (395.99 mM)
SMILES C#CC1=CC(NC2=C(C=C(OCCOCCOCCO3)C3=C4)C4=NC=N2)=CC=C1
Molecular Weight (g/mol) 391.42
Formula Weight 391.42
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.