missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICOS (CD278) Recombinant Rabbit Monoclonal Antibody (JE39-99)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556556
This item is not returnable.
View return policy
Description
ICOS (CD278) Recombinant Monoclonal Antibody for Western Blot
ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
Specifications
| ICOS (CD278) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9R1T7, Q9WVS0 | |
| ICOS | |
| Recombinant protein within human icos aa 1-100/199. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| JE39-99 | |
| Unconjugated | |
| ICOS | |
| Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 54167, 64545 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction