Learn More
Invitrogen™ ICOS (CD278) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579432
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse stomach tissue, mouse spleen tissue, mouse thymus tissue, mouse small intestine tissue, rat thymus tissue, human placenta tissue. IHC: mouse spleen tissue, mouse thymus tissue, rat spleen tissue.
ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
Specifications
| ICOS (CD278) | |
| Polyclonal | |
| Unconjugated | |
| ICOS | |
| Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29851, 54167, 64545 | |
| -20°C | |
| Lyophilized |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q9R1T7, Q9WVS0, Q9Y6W8 | |
| ICOS | |
| E. coli-derived mouse ICOS recombinant protein (Position: E21-S200). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.