missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS (CD278) Mouse anti-Human , Alexa Fluor 488, Clone: DX29, BD Biosciences™
Mouse Monoclonal Antibody
£79.09 - £215.24
Specifications
| Antigen | ICOS (CD278) |
|---|---|
| Clone | DX29 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17217794
|
BD Biosciences
567004 |
25 Tests |
£82.40 £79.09 / 25 tests Save £3.31 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17207794
|
BD Biosciences
567003 |
100 Tests |
£224.00 £215.24 / 100 tests Save £8.76 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human samples. It is validated for Flow CytometrySpecifications
| ICOS (CD278) | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Activated human T cells | |
| Primary |
| DX29 | |
| Monoclonal | |
| Mouse | |
| Human | |
| ICOS; DX-29; H4; Inducible T-cell costimulator; AILIM; CVID1 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title