missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS (CD278) Armenian Hamster, PE-CF594, Clone: C398.4A, BD Horizon™
Armenian Hamster Monoclonal Antibody
Brand: BD Biosciences 565888
This item is not returnable.
View return policy
Description
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat , Rhesus Ma samples. It is validated for Flow cytometry.
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat , Rhesus Ma samples. It is validated for Flow cytometry.
Specifications
| ICOS (CD278) | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Armenian Hamster | |
| 0.1 mg | |
| Cell Biology | |
| Human, Mouse, Rat, Rhesus Monkey | |
| Purified |
| Flow Cytometry | |
| C398.4A | |
| PE-CF594 | |
| ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115 | |
| Mouse D10.G4.1 T-cell clone | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction