missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS (CD278) Armenian Hamster, BV421, Clone: C398.4A, BD Horizon™
Armenian Hamster Monoclonal Antibody
£162.64 - £177.62
Specifications
| Antigen | ICOS (CD278) |
|---|---|
| Clone | C398.4A |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15804589
|
BD Biosciences
565887 |
25 μg |
£165.00 £162.64 / Each Save £2.36 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15894579
|
BD Biosciences
565886 |
0.1 mg |
£180.00 £177.62 / Each Save £2.38 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat, Rhesus Ma samples. It is validated for Flow cytometry.
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat, Rhesus Ma samples. It is validated for Flow cytometry.Specifications
| ICOS (CD278) | |
| 0.2mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Rhesus Monkey | |
| ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115 | |
| IgG | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. |
| C398.4A | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Armenian Hamster | |
| Cell Biology | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse D10.G4.1 T-cell clone | |
| Primary |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title