missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS Antibody (RM417), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89090
This item is not returnable.
View return policy
Description
ICOS Monoclonal specifically detects ICOS in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ICOS | |
| Monoclonal | |
| Unconjugated | |
| PBS, 50% Glycerol, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM417 | |
| Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin | |
| Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
| A peptide corresponding to the C-terminus of human ICOS | |
| 100 μL | |
| Adaptive Immunity, Immunology | |
| 29851 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction