missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS Antibody (CL14825) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26840JF525
This item is not returnable.
View return policy
Description
ICOS Monoclonal antibody specifically detects ICOS in Human samples. It is validated for Western Blot
Specifications
| ICOS | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
| This antibody was developed using a synthetic peptide derived from Q9Y6W8, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 29851 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL14825 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction