Learn More
ICOS Antibody (CL14825) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26840AF405
Product Code. 30299396
  • £582.00 / 0.10mL
Description
ICOS Monoclonal antibody specifically detects ICOS in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ICOS
Monoclonal
Alexa Fluor 405
Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
This antibody was developed using a synthetic peptide derived from Q9Y6W8, with the exact immunogen sequence remaining proprietary.
0.1 mL
Adaptive Immunity, Immunology
29851
Store at 4⪚C in the dark.
IgG1
Western Blot
CL14825
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified