missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ICOS Monoclonal antibody specifically detects ICOS in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | ICOS |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | AP-MAB0857 |
| Conjugate | Alexa Fluor 532 |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 |
| Host Species | Rat |
| Immunogen | Mouse ICOS recombinant protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?