missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS Antibody (670306), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB168H
This item is not returnable.
View return policy
Description
ICOS Monoclonal antibody specifically detects ICOS in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| ICOS | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 29851 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 670306 | |
| Flow Cytometry, CyTOF-ready | |
| Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
| NS0 mouse myeloma cell line transfected with mouse ICOS, Accession # Q9WVS0 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction